Electrode Cleaning and Storage Solutions, Oakton®, Cole Parmer
Supplier: Antyila Scientific
00653-04 00653-06
76371-710EA 45.24 CAD
76371-710 76371-712
Electrode Cleaning and Storage Solutions, Oakton®, Cole Parmer
Electrolytes Electrode Storage Solutions
Clean electrodes make clean measurements.
  • Easily remove grease, protein, and other deposits with cleaning solution
  • Make electrode last longer—store in a storage solution
Cleaning solution contains pepsin. After cleaning, keep your electrodes in proper working condition with our storage solution.