Diamond® RealSeal™ Bottles, Wide Mouth, High Density Polyethylene, Globe Scientific
Supplier: Globe Scientific
7010030 7010060 7010125 7010250 7010500 7011000 7012000 7174000 7184000
76360-258PK 56.05 CAD
76360-258 76360-260 76360-262 76360-264 76360-266 76360-268 76360-270 76360-272 76360-274
Diamond® RealSeal™ Bottles, Wide Mouth, High Density Polyethylene, Globe Scientific
Bottles Plastic Bottles
Diamond® RealSeal™ Plastic Laboratory Bottles meet the requirements of the most demanding applications. Rugged and durable, these bottles are a safe, shatterproof alternative to glass bottles for collection, storage, or shipping samples, media, and reagents.
  • Safe for freezer storage down to –100 °C
  • Resistant to most corrosives
  • Medical grade resins meet demanding requirements
  • Precision molded for consistent wall thickness and smooth surfaces
  • Leakproof, linerless cap for secure closing
  • Laboratory bottle quality and performance with savings associated with
More Product Information