Polymix® Laboratory Grinding Mills, PX-MFC 90 D, Kinematica
Supplier: Kinematica
35010023 35010033
76359-402EA 19357.16 CAD
76359-402 76359-404
Polymix® Laboratory Grinding Mills, PX-MFC 90 D, Kinematica
Mills
Grinding mill with powerful motor, which is suitable for all laboratory dry milling applications.
  • Powerful motor provides speeds up to 6000 min-1 with low noise emissions (less than 70 dB(A) at 6000 min-1 without load) due to brushless 3 phase AC motor
  • Milling chamber can be equipped with hammer grinding or blade grinding attachment (second milling chamber not required)
  • Integrated speed controller with LED display shows speed and status messages
  • Integrated overheating, overload and blocking protection means motor shuts down if overload conditions arise,
More Product Information