PIG® Spill Kit in 20-Gallon High-Visibility Container, NEW PIG
Supplier: New Pig
KIT264
76358-920EA 522.77 CAD
76358-920
PIG® Spill Kit in 20-Gallon High-Visibility Container, NEW PIG
Spill Control Materials
Hi-viz spill kit container is easily seen even in low light or from a distance to help make spill response even faster.
  • PIG® Blue Socks stop spreading spills; PIG Mat and Pillows absorb; wipers clean surfaces and hands
  • Prepacked containers are organized for easy access
  • Lime green drum is easy to locate
  • Lightweight, polypropylene drum resists chemicals and keeps contents clean and dry
  • Bold "Spill Kit" lettering for quick identification
  • Tamperproof seal helps prevent pilfering of spill response supplies
  • Temporary disposal bags with ties help make cleanup
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable