Quick-DNA/RNA Viral MagBead, Zymo Research

Supplier: Zymo Research

R2140 R2141 R2140-E R2141-E
76358-894EA 585.55 CAD
76358-894 76358-895 76358-897 76469-376 76469-378
Quick-DNA/RNA Viral MagBead, Zymo Research
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents

Quick-DNA/RNA Viral MagBead kit is designed for high-throughput purification of viral DNA and/or RNA from plasma, serum, urine, cell culture media, blood, saliva, cellular suspensions, biopsies and swab and fecal samples stored in DNA/RNA Shield (for sample collection, nucleic acid preservation and inactivation of pathogens).


  • High-throughput, magnetic-bead based purification of viral DNA and RNA from plasma, serum, urine, cell culture media, blood, saliva, cellular suspensions, swab, fecal and biopsy samples
  • High-quality DNA/RNA ready for Next-Gen sequencing, RT/qPCR, hybridization, etc
  • DNA/RNA Shield included for sample collection, inactivation and storage


The kit features a buffer system that facilitates complete viral particle lysis for efficient nucleic acid isolation. Small (> 50 nt) and large (> 200 kb) DNA and RNA are bound to magnetic beads, washed and eluted.


Isolated high-quality nucleic acids are ready for all downstream applications such as Next-Gen sequencing, hybridization-based and RT/qPCR detection.


Materials: ZR-96 MagStand, Collection Plate, 96-Well Block, Elution Plate, Cover Foil


Certifications: Meets ASTM standards. EN 342. Quick-DNA/RNA Viral MagBead – DX are CE, IVD certified.


Accessories information: ZR-96 MagStand (VWR# 76005-120)
Collection Plate (VWR# 76005-214; capacity 1.2 ml/well)
96-Well Block (VWR#s 76005-116, 76004-722, 76005-118)
Elution Plate (VWR# 76004-222; capacity 0.35 ml/well)
Cover Foil ( VWR#s 76004-236, 76005-218, 76005-220, 76005-222, 76004-234, 76005-216)


Ordering information: Equipment needed: Magnetic stand or separator, liquid handler or robotic sample processor (user provided).

Order Now

SPECIFICATIONS

Environmentally Preferable
Application
Target molecule
Refine target molecule
Sample type
Method / Format
Yield 10 µg DNA/RNA binding capacity (per 20 µl MagBinding Beads), ≥50 µl elution volume

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR