VWR® 12-Channel Reagent Reservoirs
Supplier: VWR International
76358-584 76358-586
76358-584CS 1361.85 CAD
76358-584 76358-586
VWR® 12-Channel Reagent Reservoirs
Reagent Reservoirs
These 12-channel reservoirs are made from high-quality, chemical resistant PP.
  • 12 individual channels
  • Each channel holds 5ml of solution and is individually numbered
  • Designed for complete sample withdrawal
  • Individually packaged
  • Have tabs on the bottom to ensure secure stackability
  • Includes a fitted lid
The reservoirs are ideal for all liquid handling applications. The rigid design of this product assures consistency in an automated environment. Each channel holds 5 ml of solution and is designed w/ U shaped channels to maximize complete sampleMore Product Information