Stainless Steel Baffle Assembly for Bioreactor
Supplier: Bellco Glass
1964-88610 1964-88630
76356-306EA 1329.11 CAD
76356-306 76356-308
Stainless Steel Baffle Assembly for Bioreactor
Stirrers Stirring Tools
Manufactured from 316 L stainless steel with RA32 finish or better inside and out. The assemblies attach to the headplate.
Order Now
SPECIFICATIONS
Environmentally Preferable