CORIO™ CP Heating Circulator Baths, JULABO USA, Inc.
Supplier: JULABO
9013504.22 9013504.33 9013506.22 9013506.33 9013512.22 9013512.33 9013526.22 9013526.33 9013504.22 9013504.33 9013506.22 9013506.33 9013512.22 9013512.33 9013526.22 9013526.33
76353-820EA 8285.9 CAD
76353-820 76353-822 76353-824 76353-826 76353-828 76353-830 76353-832 76353-834 CA76353-820 CA76353-822 CA76353-824 CA76353-826 CA76353-828 CA76353-830 CA76353-832 CA76353-834
CORIO™ CP Heating Circulator Baths, JULABO USA, Inc.
Circulating Thermostats Open Circulator Systems Baths with Integral Circulators
CORIO™ CP heating circulator combinations with stainless steel bath tanks for internal or external temperature applications from 20 to 200 °C.
  • Infinitely adjustable pressure pump
  • Easy to switch between internal and external circulation
  • Bright LED display
  • USB and RS232 communication ports
For internal and external temperature applications. High quality stainless steel bath tanks with bath lid and drain tap.
CORIO CP circulators feature bright LED display with push button control. A lever below the display enables simple adjustment between internalMore Product Information