Hei-VAP Expert Control Rotary Evaporators, Heidolph®
Supplier: Heidolph NA, LLC
036041020 036041021 036041022 036041023 036041024 036041025 036041026 036041027 036041028 036041029 036041030 036041031 036041032 036041033 036041034 036041035 036041036 036041037 036041038 036041039
76338-360EA 22762.59 CAD
76338-360 76338-362 76338-364 76338-366 76338-368 76338-370 76338-372 76338-374 76338-376 76338-378 76338-380 76338-382 76338-384 76338-386 76338-388 76338-390 76338-392 76338-394 76338-396 76338-398
Hei-VAP Expert Control Rotary Evaporators, Heidolph®
Evaporators Rotary Evaporators
Hei-VAP Expert Control allow to centrally control all process parameters: vacuum, cooling temperature, rotation and heating bath temperature.
  • 5" touch display for controlling the entire system including the vacuum and the cooling temperature
  • For precise monitoring, the pictogram on the display also symbolizes active functions
  • Separate knobs for quick access: the right one for direct vacuum control, the left one for the rotation
  • Setting of values either via the touch display or the separate quick access knobs
  • Direct access to two favorites on the start
More Product Information