Epredia™ Fine Pour Cassettes, Epredia
Supplier: Epredia
B851734BE B851734BL B851734GN B851734GY B851734LA B851734LG B851734LI B851734OR B851734PK B851734TN B851734WH B851734YW B851735BE B851735BL B851735GN B851735GY B851735LA B851735LG B851735LI B851735OR B851735PK B851735TN B851735WH B851735YW
76337-796CS 627.03 CAD
76337-796 76337-798 76337-800 76337-802 76337-804 76337-806 76337-808 76337-810 76337-812 76337-814 76337-816 76337-818 76337-820 76337-822 76337-824 76337-826 76337-828 76337-830 76337-832 76337-834 76337-836 76337-838 76337-840 76337-842
Epredia™ Fine Pour Cassettes, Epredia
Embedding Cassettes Biopsy Processing Cassettes
Fine pour cassettes are designed for ease of use and print clarity.
  • For very small biopsy tissue specimens
  • Variety of 12 colors
  • Comes either in plastic tubes (sleeves) or loose bulk packaging
  • Includes matching lids attached
  • Pore style: Small holes
Surface finish and reinforced print faces are engineered to optimize print quality. Tube design is precisely engineered to reduce jamming and prevent downtime.
Cassette design and flow characteristics tested to ensure efficient reagent exchange. Opening and closing forces adjusted for ease of use, whileMore Product Information