Epredia™ Cassette II Biopsy Cassettes, Epredia
Supplier: Epredia
B851730BE B851730BL B851730GN B851730GY B851730LA B851730LG B851730LI B851730OR B851730PK B851730TN B851730WH B851730YW B851738BE B851738BL B851738GN B851738GY B851738LA B851738LG B851738LI B851738OR B851738PK B851738TN B851738WH B851738YW
76337-772CS 324.52 CAD
76337-772 76337-774 76337-776 76337-778 76337-780 76337-782 76337-784 76337-786 76337-788 76337-790 76337-792 76337-794 76337-892 76337-894 76337-896 76337-898 76337-900 76337-902 76337-904 76337-906 76337-908 76337-910 76337-912 76337-914
Epredia™ Cassette II Biopsy Cassettes, Epredia
Embedding Cassettes Biopsy Processing Cassettes
Cassette II biopsy cassettes are designed for ease of use and print clarity.
  • For biopsy tissue specimens
  • Variety of 12 colors
  • Comes either in plastic tubes (sleeves) or loose bulk packaging
  • Includes matching lids
  • Pore style: Small squares
Surface finish and reinforced print faces are engineered to optimize print quality. Tube design is precisely engineered to reduce jamming and prevent downtime.
Cassette design and flow characteristics tested to ensure efficient reagent exchange. Opening and closing forces adjusted for ease of use, whileMore Product Information