Epredia™ Cassette II Slotted Cassettes, Epredia
Supplier: Epredia
B851729BE B851729BL B851729GN B851729GY B851729LA B851729LG B851729LI B851729OR B851729PK B851729TN B851729WH B851729YW B851739BE B851739BL B851739GN B851739GY B851739LA B851739LG B851739LI B851739OR B851739PK B851739TN B851739WH B851739YW
76337-748CS 324.28 CAD
76337-748 76337-750 76337-752 76337-754 76337-756 76337-758 76337-760 76337-762 76337-764 76337-766 76337-768 76337-770 76337-916 76337-918 76337-920 76337-922 76337-924 76337-926 76337-928 76337-930 76337-932 76337-934 76337-936 76337-938
Epredia™ Cassette II Slotted Cassettes, Epredia
Embedding Cassettes Biopsy Processing Cassettes
Cassette II slotted cassettes are designed for ease of use and print clarity.
  • For routine tissue specimens
  • Variety of 12 colors
  • Comes either in plastic tubes (sleeves) or loose bulk packaging
  • Includes matching lids
  • Pore style: Slotted
Surface finish and reinforced print faces are engineered to optimize print quality. Tube design is precisely engineered to reduce jamming and prevent downtime.
Cassette design and flow characteristics tested to ensure efficient reagent exchange. Opening and closing forces adjusted for ease of use, while safeguardingMore Product Information