Epredia™ HistoScreen™ Cassettes, Epredia
Supplier: Epredia
B851000BE B851000BL B851000GN B851000GY B851000LA B851000LG B851000LI B851000OR B851000PK B851000TN B851000WH B851000YW B851001BE B851001BL B851001GN B851001GY B851001LA B851001LG B851001LI B851001OR B851001PK B851001TN B851001WH B851001YW
76337-484CS 1696.11 CAD
76337-484 76337-486 76337-488 76337-490 76337-492 76337-494 76337-496 76337-498 76337-500 76337-502 76337-504 76337-506 76337-508 76337-510 76337-512 76337-514 76337-516 76337-518 76337-520 76337-522 76337-524 76337-526 76337-528 76337-530
Epredia™ HistoScreen™ Cassettes, Epredia
Embedding Cassettes Biopsy Processing Cassettes
HistoScreen™ cassette mesh screen design accommodates extremely small biopsy specimens.
  • Designed for handling small biopsies and fragile specimens
  • Unique design provides an inner chamber lined with a 'see-through' mesh
  • Eliminates the need for sponges or lens papers
  • Comes either in plastic tubes (sleeves) or loose bulk packaging
  • Variety of 12 colors with matching lids
  • Pore style: Mesh
Surface finish and reinforced print faces are engineered to optimize print quality. Tube design is precisely engineered to reduce jamming and prevent downtime.
CassetteMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable