Anti-CASB Rabbit Polyclonal Antibody

Supplier: Biosensis
R-1794-100
76334-034EA 559.08 CAD
76334-034
Anti-CASB Rabbit Polyclonal Antibody
Antibodies
Beta-casein has an important role in determination of the surface properties of the casein micelles. it is cleaved into 3 chains (casoparan, casohypotensin and antioxidant peptide). Casoparan acts as a macrophage activator, increasing the phagocytic activity of macrophages and peroxide release from macrophages. It also acts as a bradykinin-potentiating peptide. Casohypotensin acts as a bradykinin-potentiating peptide and induces hypotension in rats. Acts as a strong competitive inhibitor of endo-oligopeptidase A. Antioxidant peptide has antioxidant activity (ref: uniprot.org.).

Bovine milkMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol CASB
Environmentally Preferable
Antigen name Beta-casein
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Bovine
Host Rabbit
Gene ID CSN2
Isotype IgG
Western blot Yes
ELISA Yes
Immunogen Bovine beta-casein peptides
Form Lyophilized
Purification Affinity purified
Shipping temperature Room (ambient) temperature

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR