Anti-HSPB1 Mouse Monoclonal Antibody [clone: 6H11]
Supplier: Biosensis
M-1692-100
76334-004EA 542.49 CAD
76334-004
Anti-HSPB1 Mouse Monoclonal Antibody [clone: 6H11]
Antibodies
The heat shock proteins were discovered, as the name suggests, since they are heavily upregulated when cells are stressed by temperatures above the normal physiological range. They are expressed in unstressed cells also and have a normal function as chaperones, helping other proteins to fold correctly, and are required in much greater amounts if the cell or tissue is stressed by heat. The increased levels are generated transcriptionally under the influence of a powerful transcription factor, the heat shock factor 1 (HSF1). The different heat shock proteins were originally named based onMore Product Information