Gasket, Perfluoro, Black, 400 mm DBR Industries
Supplier: Chemglass
CG-1968-H-02
76333-492EA 5800.18 CAD
76333-492
Gasket, Perfluoro, Black, 400 mm DBR Industries
Seals Gaskets
Class VI Perfluoroelastomer with 3 tabs each spaced 120 degrees apart along the outer edge to aid in gasket alignment.
  • Gasket is for use with 400 mm Chemglass flat ground flange on Chemglass 50 and 100 L vessels
The chemically resistant perfluoroelestomer has a soft pliable sealing surface that provides improved sealing under full vacuum, as opposed to encapsulated o-ring materials and envelope gaskets.