Micro Allihn Condenser, 10/18 Joint, Chemglass
Supplier: Chemglass
CG-MR-50C1
76333-426EA 424.07 CAD
76333-426
Micro Allihn Condenser, 10/18 Joint, Chemglass
Condensers
Micro Allihn Condenser, 10/18 lower inner joint, compatible with Mettler Easymax®.
Ordering information: Supplied complete with caps with apertures and hose connections.