HemoRoamXL Mobile Blood and Plasma Refrigerators, Roemer Industries
Supplier: ROEMER INDUSTRIES, INC.
HEMOROAM8XL HEMOROAM15XL HEMOROAM25XL
76333-164EA 21174.53 CAD
76333-164 76333-226 76333-232
HemoRoamXL Mobile Blood and Plasma Refrigerators, Roemer Industries
Refrigerators
HemoRoamXL units are mobile, battery-powered refrigerators which enables the storage and transportation of large quantities of red blood cells, platelets or plasma, units have optional accessories such as battery power, wheels and data management.
  • Smart charge technology switches charge automatically between battery and electrical power plugs
  • Digital microprocessor temperature controller with built-in alarm
  • Emergency alarms if battery charge is too low
  • External temperature port ready for optional data loggers
  • Temperature uniformity of ±1.0 °C (at set point
More Product Information