VWR® Microcentrifuge Tubes for Boiling Applications
Supplier: VWR International
76332-060 76332-062 76332-064 76332-070 76332-072 76332-074 76332-076 76332-080 76414-040 76414-042 76414-044 76437-214 76437-216
76332-060CS 3244.19 CAD
76332-060 76332-062 76332-064 76332-070 76332-072 76332-074 76332-076 76332-080 76414-040 76414-042 76414-044 76437-214 76437-216
VWR® Microcentrifuge Tubes for Boiling Applications
Tubes Microcentrifuge Tubes
Polypropylene tubes feature superior clarity and a positive seal design that withstands repeated cap closures and will not pop open during boiling.
  • Graduations every 100 µl
  • Tubes fit all standard rotors
  • Certified RNase-, DNase-, and pyrogen-free
  • Certified PCR inhibitor- and human DNA-free
  • Autoclavable
  • 1.5 ml tubes are rated to 30000 ×g when properly supported in the rotor
Temperature range is from –80 to +121 °C (–112 to 250 °F).
Ordering information: Assorted colors include red, blue, green, yellow, and purple tubes.
Order Now
SPECIFICATIONS
Environmentally Preferable