Homogenizer, T 25 easy clean digital
Supplier: IKA Works
0025002561
76329-872EA 6311.14 CAD
76329-872
Homogenizer, T 25 easy clean digital
Homogenizers Benchtop Homogenizers
The T 25 easy clean digital is a high-performance dispersing instrument with electronic speed control from 3000 - 25000 rpm for working volumes of 1 - 2000 ml (H₂O).
  • No disassembly required
  • Brushless motor guarantees dust-free and silent operation
  • USB interface for control and documentation through labworldsoft®
  • Timer/counter function
  • Increased safety through tool recognition and IP-class
The S 25 dispersing elements allows the T 25 to be used in various applications- whether it is homogenizing medical samples, emulsifying cosmetics or the suspensionMore Product Information