VWR® A Balances with Calibration Certificate
Supplier: VWR International
76329-734 76329-736 76329-738 76329-740 76329-742 76329-960 76329-962 76458-518 76458-520 76458-522 76458-524 76458-526 76458-528 76458-530
76329-734EA 3462.17 CAD
76329-734 76329-736 76329-738 76329-740 76329-742 76329-960 76329-962 76458-518 76458-520 76458-522 76458-524 76458-526 76458-528 76458-530
VWR® A Balances with Calibration Certificate
Balances and Scales
The VWR® A-Series balances with calibration certificate are the answer for an organizations regulatory and compliance needs.
  • Complete calibration data reported
  • 2× RS 232, USB (A and B), and Wi-Fi communication interfaces
  • Five year warranty
  • Internal calibration
  • Easy-to-read, adjustable, backlit display
  • Tool-free assembly and diassembly of the 3-door draft shield on analytical models
The VWR® A-Series balances with calibration certificate include an ISO 17025:2017 certificate of calibration showing the full testing data using NIST-traceable weights.More Product Information