Actero™ EZ-Media Bag, FoodChek
Supplier: Foodchek
FCM-256 FCM-257 FCM-258 FCM-259 FCM-261 FCM-263
76325-092EA 4594.7 CAD
76325-092 76325-094 76325-096 76325-098 76325-100 76325-102
Actero™ EZ-Media Bag, FoodChek
Bags Media Bags
The Actero™ EZ-Media Bag contains high quality, pre-measured and sterilized enrichment media powder that greatly reduces the typical steps and glassware required in traditional media preparation.
  • Increases safety – Contains sterile enrichment medium that eliminates autoclaving or hot glassware
  • Eliminates human error – Contains pre-weighed medium, just add deionized water through the filter (ordered separately) and gently massage the bag to mix
  • Reduces time – Eliminates measuring, heating or standard consumables
  • Improves testing throughput – Quick to
More Product Information