MegaSwat & NovaPoly Mop Covers
Supplier: Micronova
QDMS-16 MS6-8IR MS6-8 QDMS-8 MS6-16NS MSAA-8NS MSAW-24NS MSAA-16NS MS1111-8 MS1111-8IR MS1111-16 MS1111-16IR MS6-8NSIR MS6-18NSIR MS6-16NSIR
68400-208EA 157.91 CAD
68400-208 89092-750 89134-958 89134-960 68400-412 89508-390 10142-782 76292-768 76323-706 76323-708 76323-710 76323-712 76443-122 76443-124 77531-060
MegaSwat & NovaPoly Mop Covers
Mops
The MegaSwat™ blade-style mop is ideal for cleaning around, under, or behind heavy process equipment or calibrated apparatus in the lab and cleanroom. Featuring a flexible polyethylene blade with a choice of covers. Lightweight and easy to use, the low-profile mop can be used for wet or dry cleaning or disinfecting in ISO 5 to 9 environments.
  • Single-ply and foam lined mops easily glide under and behind equipment
  • Lengths range from 8 to 36", and widths range from ⁵/₈ to 2"
  • Compatible with alcohol, strong disinfectants, and steam sterilization
  • Autoclavable
More Product Information