VWR® Universal Pipette Tips
Supplier: VWR International
76322-132 76322-134 76322-136 76322-138 76322-140 76322-142 76322-144 76322-146 76322-148 76322-150 76322-152 76322-154 76322-156 76323-388 76323-390 76323-392 76323-394 76323-396 76323-398 76323-400 76323-402 76323-404 76323-452 76323-454 76323-456 76322-512 76322-514 76322-516 76322-518 76322-520 76322-522 76322-524 76322-526 76322-528 76327-214 76595-790 76595-792 76595-798 76595-788 76595-794 76595-800 76595-796
76322-132CS 1257.86 CAD
76322-132 76322-134 76322-136 76322-138 76322-140 76322-142 76322-144 76322-146 76322-148 76322-150 76322-152 76322-154 76322-156 76323-388 76323-390 76323-392 76323-394 76323-396 76323-398 76323-400 76323-402 76323-404 76323-452 76323-454 76323-456 76322-512 76322-514 76322-516 76322-518 76322-520 76322-522 76322-524 76322-526 76322-528 76327-214 76595-790 76595-792 76595-798 76595-788 76595-794 76595-800 76595-796
VWR® Universal Pipette Tips
Pipette Tips
VWR® Universal low retention pipette tips repel liquid by creating a hydrophobic surface on the inside of the tip increasing your accuracy and data quality.
  • Compatible with a wide range of pipettes
  • Maximizes sample recovery with low binding resin and a fine point design
  • Improves ergonomics by reducing the force required to attach and eject a tip
  • Autoclavable at 15 psi, 122 °C for 15 minutes
  • Manufactured under an ISO 9001 quality management system
VWR® tips use a low retention resin that is up to ten times more hydrophobic than other polypropyleneMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable Waste Reducing,Sustainable Packaging Waste Reducing,Sustainable Packaging Waste Reducing,Sustainable Packaging
Environmental certification FSC-STD-40-004 V3-1 and FSC-C154485; Compared to individually racked products, CleanPak Reloads reduce plastic waste by 49% FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic FSC-STD-40-004 V3-1 and FSC-C154485; Bulk tips reduce plastic waste by 95%