VWR® Traceable® Temperature Data Loggers with TraceableGO™ Wireless Capability and Calibration
Supplier: VWR International
76321-888 76321-890 76321-892
76321-888EA 200.51 CAD
76321-888 76321-890 76321-892
VWR® Traceable® Temperature Data Loggers with TraceableGO™ Wireless Capability and Calibration
Data Loggers
Easily access collected data between destinations with Bluetooth® data loggers.
  • Connect to data via smartphone or tablet (any Bluetooth-enabled device) using a downloadable Bluetooth App
  • View and download data with free TraceableGO™ App
  • Data loggers store up to 90 alarm events
  • Readings and setup parameters easily viewable on large 2×1" LCD
  • Get accurate, reliable measurements with included NIST-traceable calibration
  • One-year warranty
  • Compatible with TraceableGO or TraceableLIVE mobile apps
  • Critical Information(Temperature, trip duration, MIN/MAX, battery
More Product Information