VWR® Syringe Pipette Tips, Evolution
Supplier: VWR International
76321-574 76321-576 76321-578 76321-580 76321-582 76321-584 76321-586 76321-588 76321-816 76321-818 76321-820 76321-822 76321-824 76321-826
76321-574PK 313.93 CAD
76321-574 76321-576 76321-578 76321-580 76321-582 76321-584 76321-586 76321-588 76321-816 76321-818 76321-820 76321-822 76321-824 76321-826
VWR® Syringe Pipette Tips, Evolution
Pipette Tips
Evolution syringe tips are compatible with most repeating pipettors on the market. Positive displacement provides precise dosing at even the smallest volumes.
  • Syringe tips come with a color coded adapter for quick identification of the correct size tip
  • Seven tip sizes dispense volumes from 1 µl to 10 ml
  • Available sterile and non sterile
All sizes of the Evolution syringe tips provide reliable and repeatable dosing for a wide range of liquids including viscous, volatile, and infectious material. Sterile tips are individually wrapped and certified to beMore Product Information