Ace-Thred™ Adapter for Swagelok® Precision Ground Tube Compression Fittings, Ace Glass Incorporated
Supplier: Ace Glass
5042-10 5042-22 5042-23 5042-24 5042-33 5042-34 5042-35 5042-53 5042-54
76318-452EA 208.51 CAD
76318-452 76318-454 76318-456 76318-468 76318-470 76318-472 76318-474 76318-476 76318-478
Ace-Thred™ Adapter for Swagelok® Precision Ground Tube Compression Fittings, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
Borosilicate glass Ace-Thred™ adapters are precision ground to accept Swagelok® type tube compression fittings. Available in #7 to #25 Ace-Thred sizes.
Order Now
SPECIFICATIONS
Environmentally Preferable