Corepoint Scientific™ White Diamond Series Glass and Solid Door Refrigerators, Horizon Scientific
Supplier: Horizon Scientific
NSWDR121WWG/0 NSWDR161WWG/0 NSWDR231WWG/0 NSWDR261WWG/0 NSWDR332WWG/0 NSWDR362WWG/0 NSWDR472WWG/0 NSWDR492WWG/0 NSWDR693WWG/0 NSWDR723WWG/0 NSWDR121WWS/0 NSWDR161WWS/0 NSWDR231WWS/0 NSWDR261WWS/0 NSWDR362WWS/0 NSWDR492WWS/0 NSWDR723WWS/0
76314-436EA 12761.19 CAD
76314-436 76314-438 76314-440 76314-442 76314-444 76314-446 76314-448 76314-450 76314-452 76314-454 76314-456 76314-458 76314-460 76314-462 76314-464 76314-466 76314-468
Corepoint Scientific™ White Diamond Series Glass and Solid Door Refrigerators, Horizon Scientific
Refrigerators
These White Diamond laboratory refrigerators offer professional cold storage for scientific and clinical settings.
  • Premium touchscreen controller
  • Advanced data logging capabilities
  • Microprocessor temperature control
  • Integrated temperature alarms
  • Digital 24-hour temperature chart
  • Keyed door lock
  • Natural hydrocarbon refrigerants
  • ENERGY STAR certified*
This line showcases elite features such as state-of-the-art touchscreen controllers, advanced alarms, data logging capabilities and unparalleled temperature control. All units run on natural, hydrocarbonMore Product Information