VWR® Vacuum Filtration Reservoir Bottles, PS
Supplier: VWR International
76311-770 76311-772 76311-774 76311-776
76311-770CS 154.38 CAD
76311-770 76311-772 76311-774 76311-776
VWR® Vacuum Filtration Reservoir Bottles, PS
Bottles Plastic Bottles
Sterile polystyrene collection reservoir bottles for solution storage.
  • DNase/RNase-free and non-pyrogenic
  • Concave design for easy handling
  • Sterilized by gamma radiation
Available in 150, 250, 500, and 1000 ml sizes.
Order Now
SPECIFICATIONS
Environmentally Preferable
Environmental certification ISO 14001/Certificate No.: 04923E00243R0M