Memmert Stainless Steel Grid Shelves
Supplier: Memmert
E28884 E20164 E20165 E28891 E20182 B41251 B38955
76311-372EA 180.27 CAD
76311-372 76311-374 76311-376 76311-534 76311-536 76311-540 76311-542
Memmert Stainless Steel Grid Shelves
Furniture Shelves
Memmert stainless steel grid shelf for use in model size 32 liters up 2140 liters.
  • Available in different sizes based on model
  • Compatible with Memmert universal ovens, climate chambers, and incubators
  • Easy to slide into place and adjust to desired height
Order Now
SPECIFICATIONS
Environmentally Preferable