Notrax® 826 Diamond Stat™ Floor Mattings, Justrite®
Supplier: NOTRAX USA, INC.
826SGC1000 826S0035BL 826R0275BL 826R0375BL 826R0475BL 826S0023BL 826S0312BL
76310-562EA 53.96 CAD
76310-562 76310-736 76310-838 76310-850 76310-852 76310-908 76310-988
Notrax® 826 Diamond Stat™ Floor Mattings, Justrite®
Floor Mats Tacky Mats
Diamond Stat™ is a dissipative/anti-static mat designed to absorb static electricity.
  • Electrically conductive PVC diamond plate top surface
  • Dense closed cell PVC foam base
  • Diamond-plate surface texture provides added traction
  • UniFusion™ bond guaranteed for the life of the mat
  • Redstop™ non-slip backing to reduce mat slippage
  • Beveled edges on all 4 sides
  • Certified by the NFSI (National Floor Safety Institute)
  • Grounding cord included, required
The mat drains static electricity from the worker, keeping sensitive circuitry from being damaged. The durableMore Product Information