Notrax® T17 Superfoam™ Floor Mattings, Justrite®
Supplier: Justrite
T17C0036BL108 T17C0048BL108 T17S0032BL T17P0032BL T17P0034BL T17S0035BL T17P0035BL T17S0036BL T17P0036BL T17S0038BL T17P0038BL T17S0033BL T17P0033BL T17S0034BL
76310-548EA 685.66 CAD
76310-548 76310-550 76310-648 76310-662 76310-694 76310-696 76310-730 76310-734 76310-762 76310-774 76310-784 76310-920 76310-948 76310-968
Notrax® T17 Superfoam™ Floor Mattings, Justrite®
Floor Mats Traction and Anti-Fatigue Floor Mats
Superfoam™ is an extremely lightweight and durable anti-fatigue foam mat designed specifically for ease of handling and ergonomic comfort.
  • Closed cell nitrile foam, does not absorb liquids
  • Durable and abrasion resistant
  • Solid for dry areas
  • Perforated drainage holes to remove liquid and debris for worker platform
  • Non-allergenic, silicone-free compound
  • 45° beveled edges on all four sides
Made from a closed cell PVC Nitrile foam blend, Superfoam™ does not absorb liquids and is highly resistant to greases, oils, animal and vegetable fats. This allows itMore Product Information