Anti-PINK1 Rabbit Polyclonal Antibody
Supplier: Rockland Immunochemical
600-401-GU5 600-401-GU5S
76310-210EA 834.24 CAD
76310-210 76310-444
Anti-PINK1 Rabbit Polyclonal Antibody
Antibodies
PINK1 (PTEN induced putative kinase 1) belongs to the protein kinase superfamily and Ser/Thr protein kinase family. PINK1 protects against mitochondrial dysfunction during cellular stress by phosphorylating mitochondrial proteins. It is involved in the clearance of damaged mitochondria via selective autophagy (mitophagy) by mediating activation and translocation of PARK2. Targets PARK2 to dysfunctional depolarized mitochondria through the phosphorylation of MFN2. Activates PARK2 in 2 steps: (1) by mediating phosphorylation at Ser-65 of PARK2 and (2) mediating phosphorylation of ubiquitin,More Product Information