Anti-PDCD4 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-965-RTU
76309-348EA 554.58 CAD
76309-348
Anti-PDCD4 Rabbit Polyclonal Antibody
Antibodies
This antibody is designed, produced, and validated as part of a collaboration between Rockland and the National Cancer Institute (NCI) and is suitable for Cancer, Immunology and Nuclear Signaling research. Programmed cell death 4 (Pdcd4) is a novel tumor supressor. Pdcd4 directly inhibits the helicase activity of eukaryotic translation initiation factor 4A (eIF4A), a component of the translation initiation complex. Pdcd4 also suppresses the transactivation of activator protein-1 (AP-1)-responsive promoters by c-Jun. Pdcd4 contains two Akt phosphorylation sites, one at Ser67 and the other atMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol PDCD4
Environmentally Preferable
Antigen name PDCD4
Antigen synonyms Death up-regulated gene protein antibody, Dug antibody, H731 antibody, Ma3 antibody, Neoplastic transformation inhibitor antibody, Neoplastic transformation inhibitor protein antibody, Nuclear antigen H731 antibody
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity
Host Rabbit
Gene ID Q53EL6
Isotype
Western blot Yes
ELISA Yes
ImmunoChemistry Yes
ImmunoPrecipitation Yes
Form Liquid (sterile filtered)
Molecular weight 52
Purification Affinity Purified
Storage buffer 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2
Shipping temperature Wet Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR