Safety Labels and Tags, White, BradyJet™ J2000, Brady
Supplier: Brady Worldwide
J20-260-7425J J20-261-7425J J20-262-7425J J20-18-7425J J20-18-2595 J20C-2250-7425J J20C-1125-7425J
76309-218RL 415.49 CAD
76309-218 76309-220 76309-222 76309-224 76309-226 76309-228 76309-230
Safety Labels and Tags, White, BradyJet™ J2000, Brady
Labels
Ideal for full-color identification and barcode applications, complete identification for a variety of areas through your laboratory including: cryovials, tubes, eppendorf tubes, conical, lab glassware, lab instructions or housekeeping and lab equipment markings.
  • White color
  • Matte finish
  • Ink jet print technology
B-7425J - matte white polypropylene with acrylic adhesive is inkjet printable. Perfect for laboratory identification such as vials, centrifuge tubes and test tubes. Service temperature of −112 to 158 °F (−80 to 70 °C).
B-2595 - generalMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable