Color Polyester Labels, Brady
Supplier: Brady Worldwide
THT-250-494-YL THT-250-494-RD THT-250-494-BL THT-250-494-OR THT-250-494-GN THT-250-494-PK THT-179-494-YL THT-179-494-RD THT-179-494-BL THT-179-494-OR THT-179-494-GN THT-179-494-PK THT-152-494-YL THT-152-494-RD THT-152-494-BL THT-152-494-OR THT-152-494-GN THT-152-494-PK B33-179-494-YL B33-179-494-RD B33-179-494-BL B33-179-494-OR B33-179-494-GN B33-179-494-PK B33-152-494-YL B33-152-494-RD 149808 B33-152-494-OR 149810 B33-152-494-PK B33-250-494-YL B33-250-494-RD B33-250-494-BL B33-250-494-OR B33-250-494-GN B33-250-494-PK
76309-090RL 128.56 CAD
76309-090 76309-092 76309-094 76309-096 76309-098 76309-100 76309-102 76309-104 76309-106 76309-108 76309-110 76309-112 76309-114 76309-116 76309-118 76309-120 76309-122 76309-124 76309-126 76309-128 76309-130 76309-132 76309-134 76309-136 76309-138 76309-140 76309-142 76309-144 76309-146 76309-148 76309-150 76309-152 76309-154 76309-156 76309-158 76309-160 76309-162 76309-164 76309-166 76309-168 76309-170 76309-172 76309-174 76309-176 76309-178 76309-180 76309-182 76309-184 76309-186 76309-188 76309-190 76309-192 76309-194 76309-196 76309-198 76309-200 76309-202 76309-204 76309-206 76309-208 76309-210 76309-212 76309-214 76309-216 76309-232 76309-234 76309-236 76309-238 76309-240 76309-242 76310-176 76310-178 76310-180 76310-182 76310-184 76310-186 76310-188 76310-190
Color Polyester Labels, Brady
Labels
Add color to reduce errors and increase efficiency with B-494 color polyester labels. The design allows users to differentiate lab samples in a snap.
  • Six color options that allow users to distinguish many different samples
  • Custom options to fit specific process and glassware needs
  • Efficient label creation by adding the Brady workstation laboratory identification software suite
Color printing below the label surface withstands the harshest lab environments, including liquid nitrogen, hot water baths, and more. Print is resistant to common lab chemicalsMore Product Information