TransferTop™ Urine Collection and Transfer Container, Globe Scientific
Supplier: Globe Scientific
3856L 3841 3852 3856TP
76208-580CS 489.45 CAD
76208-580 76308-968 76308-970 76479-380
TransferTop™ Urine Collection and Transfer Container, Globe Scientific
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Urine Sample Collection/Preparation
TransferTop™ Urine Collection and Direct Transfer Cups feature an integrated transfer mechanism for "closed system" transfer of urine samples directly from the cup to a vacuum sample tube.
  • Requires no open transferring of samples for reduced spills, improves lab hygiene, and limits exposure to dangerous samples
  • Reduces risk of contamination for improved test results
  • Directly transfers samples to a tube suitable for analysis and transport
  • Works with most vacuum urine tubes
  • Provides consistent sample transfer volumes when used with like vacuum tubes
  • Improves
More Product Information