VWR® Utility Support Uprights
Supplier: VWR International
76308-838 76308-840 76308-842 76308-864 76308-866 76308-868 76308-870 76308-872 76308-874 76308-876 76308-878 76308-880 76308-882 76308-884 76308-928 76308-930 76308-934 76308-936 76308-938 76308-940 76551-154 76550-472 76551-054 76550-998 76550-752 USUM1200-SW
76308-838EA 1472.89 CAD
76308-838 76308-840 76308-842 76308-864 76308-866 76308-868 76308-870 76308-872 76308-874 76308-876 76308-878 76308-880 76308-882 76308-884 76308-928 76308-930 76308-934 76308-936 76308-938 76308-940 76551-154 76550-472 76551-054 76550-998 76550-752 76550-494
VWR® Utility Support Uprights
Furniture Casework
USU Utility Support Uprights are the ideal solution to adding utility support above the work surface.
  • Powder coated steel
  • Attach directly to a top and allow for easy access to utilities without reducing the usable work surface area
  • Available in three width options, and can be equipped with optional shelves, horizontal channels, and dividers
  • Services can be connected via overhead service carriers, or from underneath the surface through the mounting bracket
These adjustable cable channels can be configured to suit a range of power, gas, and data needs,More Product Information