MICROCHEM® AlphaTec® 2000 Ts Plus Coveralls, Ansell
Supplier: Ansell Canada
WH20-T-92-111-02 WH20-T-92-111-03 WH20-T-92-111-04 WH20-T-92-111-05 WH20-T-92-111-06 WH20-T-92-111-07 WH20-T-92-111-08 WH20-T-92-111-09
76304-486CS 723.06 CAD
76304-486 76304-488 76304-490 76304-492 76304-494 76304-496 76304-498 76304-500
MICROCHEM® AlphaTec® 2000 Ts Plus Coveralls, Ansell
Disposable Garments
Superior breathable microporous laminate technology offering Type 4/5/6 protection.
  • Proven protective barrier to low concentration liquid chemicals
  • Moisture vapor permeable fabric ('breathable') to help reduce the risk of heat stress
  • Optimized body fit, improving wearer comfort and safety
  • Anti-static
  • Ultra low linting, reduced risk of contamination in critical areas
  • Silicone-free, critical in spray painting applications
  • Tunnelled elasticated hood, wrists, and ankles
The MICROCHEM® by AlphaTec® 2000 Ts Plus coveralls provide both protection and comfort,More Product Information