Dry Bath Heaters with Touch Screen, 230 V
Supplier: Boekel
115001-2 115002-2 115004-2
76304-424EA 1177.81 CAD
76304-424 76304-428 76304-506
Dry Bath Heaters with Touch Screen, 230 V
Block Heaters Block Heater/Coolers
Boekel scientific's dry block heaters are advanced, configurable and easy-to-use.
  • 230 V
  • Easily viewable touch screen
  • Save up to 25 programs for rapid sample processing
  • Slim design to conserve bench space
  • Wide temperature range to fit many applications
  • Settable timer up to 99 hours or continuous
The Boekel scientific block heaters line is designed to accommodate the most basic sample incubation procedures as well as multi-step processes and thermal cycling procedures. The slim one, two and four block design allows for more bench space and aMore Product Information