Human Recombinant MMP-1 (from E. coli)
Supplier: Peprotech
420-01-50UG
76303-880EA 1524.87 CAD
76303-880
Human Recombinant MMP-1 (from E. coli)
Proteins and Peptides
Matrix metalloproteinases (MMPs) are a family of endoproteases that require zinc and calcium for expressing catalytic activity. These enzymes play a central role in the maintenance and remodeling of the extracellular matrix. Elevated expression of their activity, caused either by up-regulation of their expression or down-regulation of their cognate inhibitors, has been implicated in various degenerative disorders, including arthritis, cardiovascular disease, skeletal growth-plate disorders, and cancer metastasis. MMP-1 is a secreted collagenase with specificity toward Type I, II, III, VII, and X collagens. Recombinant Human MMP-1 is a 42.7 kDa protein containing the entire catalytic N-terminal domain and the C-terminal domain, which is involved in substrate specificity, and in binding TIMP-1.