KleenGuard™ Maverick™ Safety Glasses, Kimberly-Clark Professional®
Supplier: Ansell Healthcare
49309 49311 49312 49301
76300-904CS 285.91 CAD
76300-904 76300-906 76300-908 76300-910
KleenGuard™ Maverick™ Safety Glasses, Kimberly-Clark Professional®
Safety Glasses
Maverick™ eye protection combines a modern, sleek, and trendy design with reliable comfort and safety features.
  • Modern, sleek, stylish, and lightweight design employees are happy to wear
  • Integrated browguard and side shields help the user feel confident they are protected
  • Comfort touch points on nose bridge and temples prevent slippage and reduce pressure
  • Polycarbonate lenses provide 99.9% UVA/UVB/UVC protection
  • Premium Anti-Fog and Premium Anti-Scratch lens coatings for better performance and longer life
  • Eligible for recycling via RightCycle by Kimberly-
More Product Information