EcoSense® EC300M-Conductivity, TDS, Salinity, Temperature Instrument with Extended Memory, YSI
Supplier: YSI
601034 601035 601036 601037
76299-982EA 965.11 CAD
76299-982 76299-984 76299-986 76299-988
EcoSense® EC300M-Conductivity, TDS, Salinity, Temperature Instrument with Extended Memory, YSI
Electrochemistry Meters Conductivity Meters
The EC300M offers convenient measurement of conductivity, specific conductance, salinity, total dissolved solids, and temperature in many field applications. Compared to the EcoSense® EC300A the EC300M features extended memory and simple data transfer with a micro-USB port.
  • Holds 250 data sets; no need to write down data
  • Waterproof micro-USB port for transferring data to PC; no software required
  • Real-time clock for date/time stamp of saved data
  • User-selectable recalibration prompt provides reminder to calibrate
  • Automatic temperature compensation
  • Super-
More Product Information