TSX Series High-Performance Chromatography Refrigerators, Thermo Scientific
Supplier: Thermo Fisher Scientific
TSX4505CA TSX4505CD TSX5005CA TSX5005CD TSX2305CA TSX2305CD TSX3005CA TSX3005CD
76299-964EA 24335.11 CAD
76299-964 76299-966 76299-968 76299-970 76299-972 76299-974 76299-976 76299-978 76299-980 76320-224 76320-298 76320-300
TSX Series High-Performance Chromatography Refrigerators, Thermo Scientific
Refrigerators
The TSX high-performance chromatography refrigerators are designed for a variety of applications requiring close temperature control, full access to chromatography instrumentation, and easy set-up of instrumentation and apparatus within the chamber. The TSX chromatography series features industry-leading safety features to ensure maximum protection.
  • Forced-air circulation for temperature uniformity at all shelf levels and Heat-free defrost for maximum temperature uniformity
  • Quiet operation at just 52 dBA
  • Two 2" (5 cm) side wall access ports with insulated
More Product Information