Miller® TurboLite™ MAX Smooth Edge Personal Fall Limiters, Honeywell Safety
Supplier: Honeywell Safety Products
MFLEW-11/12FT MFLEW-11/6FT MFLEW-11/9FT MFLEW-12/12FT MFLEW-12/6FT MFLEW-12/9FT MFLEW-18/12FT MFLEW-18/6FT MFLEW-18/9FT MFLEW2-11/12FT MFLEW2-12/9FT MFLEW2-18/12FT MFLEW2-18/6FT MFLEW2-3/12FT MFLEW2-3/6FT MFLEW2-3/9FT MFLEW2-11/6FT MFLEW2-11/9FT MFLEW2-12/12FT MFLEW2-12/6FT MFLEW2-4/12FT MFLEW2-4/6FT MFLEW2-4/9FT MFLEW-3/12FT MFLEW-3/9FT MFLEW-4/12FT MFLEW-4/6FT MFLEW-4/9FT
76299-714EA 804.4 CAD
76299-714 76299-716 76299-718 76299-720 76299-722 76299-724 76299-726 76299-728 76299-730 76299-732 76299-734 76299-736 76299-738 76299-740 76299-742 76299-744 76299-746 76299-748 76299-750 76299-752 76299-754 76299-756 76299-758 76299-760 76299-762 76299-764 76299-766 76299-768 76299-770 76299-772
Miller® TurboLite™ MAX Smooth Edge Personal Fall Limiters, Honeywell Safety
Fall Protection Fall Lanyards and Ropes
Web lifeline for smooth edge applications.
  • Designed for use in smooth edge applications with radius ≥ 0.060" (1.5 mm)
  • Honeywell Spectra® webbing lifeline provides outstanding chemical, abrasion, and cut resistance
  • Available in lengths of 6, 9 and 12'
  • Single and twin configurations
  • Variety of lifeline connector options
Miller® TurboLite™ Personal Fall Limiters (PFLs) are designed for performance in edge applications, offering versatility, enhanced user mobility, and maximum safety. These devices maximize versatility for virtually any short-rangeMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable