VertiKlean® MAX™ Mopping Systems, Contec®
Supplier: Contec
2691A VKMX0175 VKMX0325 VKMX0375 VKMX0125 2652SS 2653SS VKSE0100 VKSE0150 VKSE0300 VKSE0350 2652LF 2651LF
33502-721EA 278.13 CAD
33502-721 33503-126 89074-000 10119-376 10119-380 10119-382 10171-154 10017-722 10816-272 10816-274 76169-352 76169-354 76169-356 76199-898 76199-908 76299-418 76449-910
VertiKlean® MAX™ Mopping Systems, Contec®
Mops
VertiKlean® MAX™ Mop was designed for applying disinfectants to both horizontal and vertical surfaces, and is ideal for general cleaning of ceilings, walls, and floors in the most critical environments. This durable, lightweight mop features a single layer hydrophilic polyurethane foam laminated to polyester microfiber fabric, backed by a semi-rigid thermoformed plate, which easily snaps onto a standard VertiKlean® mop frame.
  • Hydrophilic polyurethane foam and knitted polyester mop head retains liquids
  • When the mop is applied to a surface, the liquid is
More Product Information