MemBrite™ Fix Cell Surface Staining Kits, Biotium
Supplier: Biotium
30092-T 30092 30093-T 30093 30094-T 30094 30095-T 30095 30096-T 30096 30097-T 30097 30098-T 30098 30099-T 30099
76294-116EA 455.07 CAD
76294-116 76294-126 76294-128 76294-130 76294-132 76294-134 76294-136 76294-138 76294-140 76294-142 76294-144 76294-146 76294-148 76294-150 76294-152 76294-154
MemBrite™ Fix Cell Surface Staining Kits, Biotium
Cell Strainers
MemBrite™ Fix cell surface staining kits are designed for covalently staining the surface of live cells to conveniently visualize cell boundaries in immunofluorescence experiments.
  • Rapid, uniform live cell surface staining
  • Covalent fluorescent labeling can be fixed and permeabilized for IF staining
  • Stain mammalian cells, yeast, or gram-positive bacteria
  • Choose from 12 bright and photostable dye colors from blue to near-IR
  • MemBrite™ Fix-ST dyes for super-resolution STORM imaging
These stains are novel reactive fluorescent dyes stains that reactMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable