Cork Stoppers, United Scientific Supplies
Supplier: United Scientific Supplies
CST000-VWR CST00-VWR CST0-VWR CST1-VWR CST2-VWR CST3-VWR CST4-VWR CST5-VWR CST6-VWR CST7-VWR CST8-VWR CST9-VWR CST10-VWR CST11-VWR CST12-VWR CST13-VWR CST14-VWR CST15-VWR CST16-VWR CST17-VWR CST18-VWR CST19-VWR CST20-VWR
76293-818PK 9.85 CAD
76293-818 76293-820 76293-822 76293-824 76293-826 76293-828 76293-830 76293-832 76293-834 76293-836 76293-838 76293-840 76293-842 76293-844 76293-846 76293-848 76293-850 76293-852 76293-854 76293-856 76293-858 76293-860 76293-862
Cork Stoppers, United Scientific Supplies
Stoppers Cork Stoppers
Cork stoppers function as a closure for bottles, vials, laboratory vessels and other openings.
  • Regular length taper
  • Assorted sizes
These grade XXX tapered cork stoppers are an excellent choice for general laboratory use. They are effective for both liquid and solid mediums and offer light weight, resilience, and compressibility. Chemically inert and moisture-resistant.
Packaging: Sold as packs of 100.
Order Now
SPECIFICATIONS
Environmentally Preferable