Lattice Lab Rod Connector, United Scientific Supplies
Supplier: United Scientific Supplies
LLSRC2-VWR
76293-590EA 27.32 CAD
76293-590
Lattice Lab Rod Connector, United Scientific Supplies
Supports Flask Supports
This aluminum connector is used to attach two aluminum rods together.
  • 12.9 mm inner diameter
These aluminum connectors help join two aluminum lattice rods together securely.
Delivery information: Includes two set screws and an Allen key.