Nickel Crucibles, United Scientific Supplies
Supplier: United Scientific Supplies
NCR025-VWR
76293-528EA 38.05 CAD
76293-528
Nickel Crucibles, United Scientific Supplies
Crucibles
Unibody construction from sheet nickel, polished inside and out.
  • Polished inside and out
  • Includes a cover
These nickel crucibles can withstand temperatures up to 600 °C.
Delivery information: Covers included.